2,4-Dihydroxybenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: beta-Resorcylic acid

D0568-25G
TCD0568-025GEA 74.26 CAD
TCD0568-025G
2,4-Dihydroxybenzoic acid ≥98.0%
2,4-Dihydroxybenzoic acid

CAS Number: 89-86-1
MDL Number: MFCD00002451
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 218
Flash Point (°C): 149


Other Product Names / Synonyms: beta-Resorcylic Acid

Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 390
Melting Pt: 218 °C
Density: 1.54 g/cm³ (20 °C)
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00002451
CAS Number: 89-86-1
EINECS: 201-946-9
Merck Index: 12,08325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbc5ee3820d0160bd20d48dc2cfcf55f44f5ae15&vid=f768d0d2be12e4beca3e0ca27af61123a5a6f6c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR