Dicloran (2,6-dichloro-4-nitroaniline) ≥97.0%

Supplier: TCI America
Danger

D0385-500G D0385-25G
TCD0385-500GEA 557.74 CAD
TCD0385-500G TCD0385-25G
Dicloran (2,6-dichloro-4-nitroaniline) ≥97.0%
Dicloran (2,6-Dichloro-4-nitroaniline)

CAS Number: 99-30-9
MDL Number: MFCD00007677
Molecular Formula: C6H4Cl2N2O2
Molecular Weight: 207.01
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 193

Formula: C₆H₄Cl₂N₂O₂
Boiling Pt: 130 °C
Melting Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00007677
CAS Number: 99-30-9
EINECS: 202-746-4
UN: 2237
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae4d6db11b43cd55cec909fd56926c1ae627f4a2&vid=8d03d90b8e258967ba1b2a6b511298e696781d7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR