3,4-Dichlorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0340-25G
TCD0340-500GEA 954.07 CAD
TCD0340-500G TCD0340-025G
3,4-Dichlorobenzoic acid ≥98.0% (by GC, titration analysis)
3,4-Dichlorobenzoic acid

CAS Number: 51-44-5
MDL Number: MFCD00002492
Molecular Formula: C7H4Cl2O2
Molecular Weight: 191.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 205

Formula: C₇H₄Cl₂O₂
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00002492
CAS Number: 51-44-5
EINECS: 200-099-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52b1f4382678f40bc6a158d79e630816c746e979&vid=e5eee97073630620802aeaf5258024bb4f29ffb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR