2,6-Dichloroaniline ≥99.0%

Supplier: TCI America
Danger

D0323-25G D0323-100G
TCD0323-25GEA 56.88 CAD
TCD0323-25G TCD0323-100G
2,6-Dichloroaniline ≥99.0%
2,6-Dichloroaniline

CAS Number: 608-31-1
MDL Number: MFCD00007675
Molecular Formula: C6H5Cl2N
Molecular Weight: 162.01
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 228
Melting point (°C): 38
Flash Point (°C): 118

Formula: C₆H₅Cl₂N
Boiling Pt: 228 °C
Melting Pt: 38 °C
Density: 1.526 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00007675
CAS Number: 608-31-1
EINECS: 210-160-5
UN: 3442
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c37b1ad4fb12801031a0f9a1acaddf0eb72c9f93&vid=caf7d2950d98a60e57162b19ea33368e020c2047&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR