1,3-Di-4-piperidylpropane ≥97.0%

Supplier: TCI America

D0293-25G
TCD0293-500GEA 618.21 CAD
TCD0293-500G TCD0293-025G
1,3-Di-4-piperidylpropane ≥97.0%
1,3-Di-4-piperidylpropane

CAS Number: 16898-52-5
MDL Number: MFCD00038013
Molecular Formula: C13H26N2
Molecular Weight: 210.37
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 66
Flash Point (°C): 137


Other Product Names / Synonyms: 1,3-Bis(4-piperidyl)propane, 4,4'-Trimethylenedipiperidine

Formula: C₁₃H₂₆N₂
Melting Pt: 66 °C
Flash Pt: 137 °C
MDL Number: MFCD00038013
CAS Number: 16898-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=449f8ec387c414ae75d3aa92719e71a5fe57d06b&vid=449f8ec387c414ae75d3aa92719e71a5fe57d06b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR