3,5-Dibromo-4-hydroxybenzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0189-25G
TCD0189-025GEA 162.74 CAD
TCD0189-025G
3,5-Dibromo-4-hydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
3,5-Dibromo-4-hydroxybenzaldehyde

CAS Number: 2973-77-5
MDL Number: MFCD00016980
Molecular Formula: C7H4Br2O2
Molecular Weight: 279.92
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 184

Formula: C₇H₄Br₂O₂
Melting Pt: 184 °C
MDL Number: MFCD00016980
CAS Number: 2973-77-5
EINECS: 221-017-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44c28d768e19b3f7e017db06e709249f7f5ffe58&vid=44c28d768e19b3f7e017db06e709249f7f5ffe58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR