4-Nitro-m-phenylenediamine ≥98.0%

Supplier: TCI America
Warning

D0104-25G
TCD0104-025GEA 99.54 CAD
TCD0104-025G
4-Nitro-m-phenylenediamine ≥98.0%
4-Nitro-m-phenylenediamine

CAS Number: 5131-58-8
MDL Number: MFCD00025289
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 160


Other Product Names / Synonyms: 1,3-Diamino-4-nitrobenzene

Formula: C₆H₇N₃O₂
Melting Pt: 160 °C
Density: 1.552 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00025289
CAS Number: 5131-58-8
EINECS: 225-876-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e72295297645463d64a8ef593cc2014ab5c1e8a6&vid=e72295297645463d64a8ef593cc2014ab5c1e8a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR