4-Nitro-o-phenylenediamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0103-25G D0103-100G
TCD0103-025GEA 40.2 CAD
TCD0103-025G TCD0103-100G
4-Nitro-o-phenylenediamine ≥97.0% (by GC, titration analysis)
4-Nitro-o-phenylenediamine

CAS Number: 99-56-9
MDL Number: MFCD00007724
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 201


Other Product Names / Synonyms: 1,2-Diamino-4-nitrobenzene

Formula: C₆H₇N₃O₂
Melting Pt: 201 °C
MDL Number: MFCD00007724
CAS Number: 99-56-9
EINECS: 202-766-3
Merck Index: 12,06720
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ff49049efe1aca2b899197c2a5c902126b6aff5&vid=2ff49049efe1aca2b899197c2a5c902126b6aff5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR