1,8-Diaminonaphthalene ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D0102-25G D0102-500G D0102-100G
TCD0102-025GEA 175.38 CAD
TCD0102-025G TCD0102-500G TCD0102-100G
1,8-Diaminonaphthalene ≥98.0% (by titrimetric analysis)
1,8-Diaminonaphthalene

CAS Number: 479-27-6
MDL Number: MFCD00004033
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 205
Melting point (°C): 65


Other Product Names / Synonyms: 1,8-Naphthalenediamine

Formula: C₁₀H₁₀N₂
Boiling Pt: 205 °C
Melting Pt: 65 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00004033
CAS Number: 479-27-6
EINECS: 207-529-8
UN: 3259
ADR: 8,III
Merck Index: 12,06459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22a7dd73ca0dfb767c8f65b86ef444ff7d78c29d&vid=22a7dd73ca0dfb767c8f65b86ef444ff7d78c29d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR