4,4'-Diaminodiphenylamine sulfate Hydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D0041-500G D0041-25G
TCD0041-500GEA 589.34 CAD
TCD0041-500G TCD0041-025G
4,4'-Diaminodiphenylamine sulfate Hydrate ≥97.0% (by titrimetric analysis)
4,4'-Diaminodiphenylamine sulfate Hydrate

CAS Number: 53760-27-3
MDL Number: MFCD00013113
Molecular Formula: C12H13N3
Molecular Weight: 297.33
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 300
Flash Point (°C): 410


Product Categories: Materials Chemistry, Monomers, Diphenylamines (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(4-aminophenyl)amine Sulfate, 4,4'-Iminodianiline Sulfate

Formula: C₁₂H₁₃N₃
Melting Pt: 300 °C
Flash Pt: 410 °C
MDL Number: MFCD00013113
CAS Number: 53760-27-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71a57c43cfbf29707965a6481e726d1c7fe58116&vid=71a57c43cfbf29707965a6481e726d1c7fe58116&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR