Sodium dehydracetate monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Sodium 3-Acetyl-4-hydroxy-6-methyl-2-oxo-4(2H)-pyranolate, Sodium Dehydroacetate monohydrate, Sodium dehydroacetate monohydrate, Dehydroacetic acid sodium salt monohydrate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate monohydrate, 3-Acetyl-4-hydroxy-6-methyl-2-pyrone sodium salt, Sodium Dehydroacetate Monohydrate, Dehydracetic acid sodium salt monohydrate

D0040-25G
TCD0040-025GEA 44.24 CAD
TCD0040-025G TCD0040-500G
Sodium dehydracetate monohydrate ≥98.0% (by HPLC, titration analysis)
Sodium dehydracetate monohydrate

CAS Number: 64039-28-7
MDL Number: MFCD00040583
Molecular Formula: C8H8O4
Molecular Weight: 190.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 3-Acetyl-4-hydroxy-6-methyl-2-pyrone Sodium Salt, Sodium 3-Acetyl-4-hydroxy-6-methyl-2-oxo-4(2H)-pyranolate, Dehydroacetic Acid Sodium Salt

Formula: C₈H₇NaO₄·1H₂O
MW: 208.15 g/mol
MDL Number: MFCD00040583
CAS Number: 64039-28-7
EINECS: 224-580-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60daf6300eb5f27d09a21aae7632f0be4aef645b&vid=60daf6300eb5f27d09a21aae7632f0be4aef645b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR