Didecyl ether ≥95.0%

Supplier: TCI America

D0035-25ML
TCD0035-025MLEA 327.06 CAD
TCD0035-025ML
Didecyl ether ≥95.0%
Didecyl ether

CAS Number: 2456-28-2
MDL Number: MFCD00026539
Molecular Formula: C20H42O
Molecular Weight: 298.56
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Didecyl Ether

Formula: C₂₀H₄₂O
Boiling Pt: 196 °C
Density: 0.82 (20 °C)
MDL Number: MFCD00026539
CAS Number: 2456-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8017242ebb92ebb26c2acd5ac7682927482c1ff1&vid=8017242ebb92ebb26c2acd5ac7682927482c1ff1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR