5-Chloro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3367-1G C3367-200MG
TCC3367-1GEA 255.96 CAD
TCC3367-1G TCC3367-200MG
5-Chloro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
5-Chloro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
MDL Number: MFCD06798284
CAS Number: 865186-94-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR