4-Chloro-5'-fluoro-2'-hydroxybenzophenone ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4'-Chloro-5-fluoro-2-hydroxybenzophenone, (4-Chlorophenyl)(5-fluoro-2-hydroxyphenyl)methanone

C3338-1G C3338-5G
TCC3338-1GEA 216.46 CAD
TCC3338-1G TCC3338-5G
4-Chloro-5'-fluoro-2'-hydroxybenzophenone ≥97.0% (by GC)
4-Chloro-5'-fluoro-2'-hydroxybenzophenone
Formula: C₁₃H₈ClFO₂
MW: 250.6500 g/mol
MDL Number: MFCD00040948
CAS Number: 62433-26-5
EINECS: 263-539-2

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR