Chloro[(1,2,3,4,5,6-eta)-2,2'',4,4'',6,6''-hexamethyl[1,1':3',1''-terphenyl]-2'-thiolato-kappaS][tris(4-fluorophenyl)phosphine-kappaP]ruthenium(II)

Supplier: TCI America

Synonyms: [(DmpSR')RuCl(P-(4-Fluorophenyl)3)]

C3328-100MG
TCC3328-100MGEA 475.58 CAD
TCC3328-100MG
Chloro[(1,2,3,4,5,6-eta)-2,2'',4,4'',6,6''-hexamethyl[1,1':3',1''-terphenyl]-2'-thiolato-kappaS][tris(4-fluorophenyl)phosphine-kappaP]ruthenium(II)
Chloro[(1,2,3,4,5,6-eta)-2,2'',4,4'',6,6''-hexamethyl[1,1':3',1''-terphenyl]-2'-thiolato-kappaS][tris(4-fluorophenyl)phosphine-kappaP]ruthenium(II)
Formula: C₄₂H₃₇ClF₃PRuS
MW: 798.31 g/mol
CAS Number: 1420299-84-8

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Elemental analysis (Carbon) 60.50 to 65.50 %
Elemental analysis (Hydrogen) 4.40 to 5.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR