2-Chloro-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(6-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloropyridine-5-boronic acid Pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

C3136-1G
TCC3136-1GEA 64.78 CAD
TCC3136-1G TCC3136-5G
2-Chloro-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Chloro-5-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
MW: 239.51
MDL Number: MFCD03412793
CAS Number: 444120-94-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR