3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C3110-1G C3110-5G
TCC3110-1GEA 116.92 CAD
TCC3110-1G TCC3110-5G
3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride ≥97.0% (by titrimetric analysis)
3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic acid 1,4:2,3-Dianhydride
Formula: C₁₀H₈O₆
MDL Number: MFCD22373052
CAS Number: 6053-46-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Methanolysis Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR