(3-Chloropropyl)diethoxy(methyl)silane 98,0 GC_ASSAY_METHOD

Supplier: TCI America
Warning

C3092-25ML C3092-5ML
TCC3092-25MLEA 131.14 CAD
TCC3092-25ML TCC3092-5ML
(3-Chloropropyl)diethoxy(methyl)silane 98,0 GC_ASSAY_METHOD
(3-Chloropropyl)diethoxy(methyl)silane
Formula: C₈H₁₉ClO₂Si
MW: 210.77 g/mol
Boiling Pt: 109°C
MDL Number: MFCD05663910
CAS Number: 13501-76-3
EINECS: 236-828-6

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR