2-Chloro-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3077-1G C3077-5G
TCC3077-1GEA 53.72 CAD
TCC3077-1G TCC3077-5G
2-Chloro-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-4-methoxybenzoic acid
Formula: C₈H₇ClO₃
MDL Number: MFCD00085943
CAS Number: 21971-21-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 210.0 to 214.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR