(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C3005-25G C3005-5G
TCC3005-25GEA 265.44 CAD
TCC3005-25G TCC3005-5G
(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate ≥98.0% (by HPLC, total nitrogen)
(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate

CAS Number: 34079-31-7
MDL Number: MFCD00020830
Molecular Formula: C13H16N2O3
Molecular Weight: 248.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 92
Specific rotation [a]20/D: -97 deg (C=0.5, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-prolinamide, N-Benzyloxycarbonyl-L-prolinamide

Formula: C₁₃H₁₆N₂O₃
Melting Pt: 92 °C
MDL Number: MFCD00020830
CAS Number: 34079-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8832aae08a3c1373daf40e3f0629b8be4f18f4d4&vid=8832aae08a3c1373daf40e3f0629b8be4f18f4d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR