3-(9H-Carbazol-9-yl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

C2967-1G C2967-5G
TCC2967-1GEA 60.04 CAD
TCC2967-1G TCC2967-5G
3-(9H-Carbazol-9-yl)phenylboronic acid (contains varying amounts of Anhydride)
3-(9H-Carbazol-9-yl)phenylboronic acid

CAS Number: 864377-33-3
MDL Number: MFCD18072506
Molecular Formula: C18H14BNO2
Molecular Weight: 287.13
Form: Crystal
Color: White


Other Product Names / Synonyms: 3-(9H-Carbazol-9-yl)benzeneboronic Acid

Formula: C₁₈H₁₄BNO₂
MDL Number: MFCD18072506
CAS Number: 864377-33-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c44f00b4a65889efd9f1e143d2b82df2af512ee&vid=8c44f00b4a65889efd9f1e143d2b82df2af512ee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR