Cloquintocet mexyl ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2682-1G
TCC2682-1GEA 74.26 CAD
TCC2682-1G
Cloquintocet mexyl ≥98.0% (by HPLC, titration analysis)
Cloquintocet mexyl

CAS Number: 99607-70-2
MDL Number: MFCD01632329
Molecular Formula: C18H22ClNO3
Molecular Weight: 335.83
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 70


Product Categories: Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: 2-Heptyl 2-[(5-Chloroquinolin-8-yl)oxy]acetate, 2-[(5-Chloroquinolin-8-yl)oxy]acetic Acid 2-Heptyl Ester

Formula: C₁₈H₂₂ClNO₃
Melting Pt: 70 °C
MDL Number: MFCD01632329
CAS Number: 99607-70-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f3a2f8e8875e05468b26e6bc935e87e9add486c&vid=0f3a2f8e8875e05468b26e6bc935e87e9add486c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR