3-Cyano-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 3-Cyano-4-fluorobenzeneboronic acid

C2676-1G
TCC2676-1GEA 80.58 CAD
TCC2676-1G
3-Cyano-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Cyano-4-fluorophenylboronic acid

CAS Number: 214210-21-6
MDL Number: MFCD03095130
Molecular Formula: C7H5BFNO2
Molecular Weight: 164.93
Form: Crystal
Color: White


Other Product Names / Synonyms: 3-Cyano-4-fluorobenzeneboronic Acid

Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
MDL Number: MFCD03095130
CAS Number: 214210-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR