5-Chloro-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

C2645-5G
TCC2645-5GEA 93.22 CAD
TCC2645-5G
5-Chloro-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
5-Chloro-2-fluorophenylboronic acid

CAS Number: 352535-83-2
MDL Number: MFCD05664225
Molecular Formula: C6H5BClFO2
Molecular Weight: 174.36
Form: Crystal
Color: White


Other Product Names / Synonyms: 5-Chloro-2-fluorobenzeneboronic Acid

Formula: C₆H₅BClFO₂
MDL Number: MFCD05664225
CAS Number: 352535-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=184290a0765f44e940a645cdf3468fc26aae5fcc&vid=184290a0765f44e940a645cdf3468fc26aae5fcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR