Copper(II) on chitosan (1,3 mmol/g Cu) (1.3 mmol/g Cu)

Supplier: TCI America

C2637-5G
TCC2637-5GEA 330.22 CAD
TCC2637-5G
Copper(II) on chitosan (1,3 mmol/g Cu) (1.3 mmol/g Cu)
Copper(II) on chitosan

Form: Crystals
Color: Light Blue


Application: Copper (II) on Chitosan, a reagent for selective discrimination between 2-hydroxy and 2-deoxy sugars; an alternative test reagent for Fehling and Benedict reagents


Copper (II) on chitosan is a chitosan-supported copper (II) (about 1.3 mmol/g) developed by Inoue et al. as a reagent for selective detection of reducible organic compounds. The Cu (II) on chitosan can selectively detect reducible organic compounds bearing structures changeable to ene-diolate (e.g. glucose, ribose) in basic aqueous solution with the formation of copper (I) oxide (Cu2O) as a signal of the reduction by the ene-diolate.

Order Now

Specification Test Results

Loading 1.2 to 1.5 mmol/g
Functionality test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR