2-Chloroquinoline-3-carbaldehyde ≥98.0%

Supplier: TCI America

C2289-1G C2289-5G
TCC2289-1GEA 33.18 CAD
TCC2289-1G TCC2289-5G
2-Chloroquinoline-3-carbaldehyde ≥98.0%
2-Chloroquinoline-3-carbaldehyde

CAS Number: 73568-25-9
MDL Number: MFCD00130079
Molecular Formula: C10H6ClNO
Molecular Weight: 191.61
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 150

Formula: C₁₀H₆ClNO
Melting Pt: 150 °C
MDL Number: MFCD00130079
CAS Number: 73568-25-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3f3ccdeba4bd429fcbc7e0a1dceb2eda9bd8aa8&vid=b3f3ccdeba4bd429fcbc7e0a1dceb2eda9bd8aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR