1-Cbz-piperazine ≥95.0% (by GC, titration analysis)

Supplier: TCI America

C2178-25G C2178-5G
TCC2178-25GEA 502.44 CAD
TCC2178-25G TCC2178-5G
1-Cbz-piperazine ≥95.0% (by GC, titration analysis)
1-Cbz-piperazine

CAS Number: 31166-44-6
MDL Number: MFCD00274317
Molecular Formula: C12H16N2O2
Molecular Weight: 220.27
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 110
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 1-Cbz-piperazine, Benzyl Piperazine-1-carboxylate, Piperazine-1-carboxylic Acid Benzyl Ester

Formula: C₁₂H₁₆N₂O₂
Boiling Pt: 161 °C
Density: 1.15 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00274317
CAS Number: 31166-44-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aac82bf3978dbcd382018e1f86f6a21533a1ef&vid=e5aac82bf3978dbcd382018e1f86f6a21533a1ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR