2-Chloro-5-methylpyridine ≥97.0%

Supplier: TCI America
Warning

C1999-5G
TCC1999-5GEA 52.14 CAD
TCC1999-5G
2-Chloro-5-methylpyridine ≥97.0%
2-Chloro-5-methylpyridine

CAS Number: 18368-64-4
Molecular Formula: C6H6ClN
Molecular Weight: 127.57
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 56
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 6-Chloro-3-picoline

Formula: C₆H₆ClN
Boiling Pt: 56 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00792460
CAS Number: 18368-64-4
EINECS: 418-050-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=064ac915389792d71450b0918c4c3f79cfa08f0e&vid=064ac915389792d71450b0918c4c3f79cfa08f0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR