Chloromethyl cyclohexyl ether ≥96.0%

Supplier: TCI America

Synonyms: (Chloromethoxy)cyclohexane

C1947-25G
TCC1947-25GEA 687.3 CAD
TCC1947-25G
Chloromethyl cyclohexyl ether ≥96.0%
Chloromethyl cyclohexyl ether

CAS Number: 3587-62-0
Molecular Formula: C7H13ClO
Molecular Weight: 148.63
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Specific Gravity (20/20): 1.06

Formula: C₇H₁₃ClO
Boiling Pt: 185 °C
Density: 1.06 (20 °C)
MDL Number: MFCD10566884
CAS Number: 3587-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6c884b3fbe6bbbfeb529d4f394b552f68e9d883&vid=f6c884b3fbe6bbbfeb529d4f394b552f68e9d883&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR