6-Chloropicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1896-1G C1896-5G
TCC1896-1GEA 64.78 CAD
TCC1896-1G TCC1896-5G
6-Chloropicolinic acid ≥98.0% (by GC, titration analysis)
6-Chloropicolinic acid

CAS Number: 4684-94-0
MDL Number: MFCD00155390
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 188
Lambda max.: 294 nm (EtOH)


Other Product Names / Synonyms: 6-Chloropicolinic Acid

Formula: C₆H₄ClNO₂
Melting Pt: 188 °C
MDL Number: MFCD00155390
CAS Number: 4684-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94d8900835afc7998559e303bc680c5f301944f5&vid=94d8900835afc7998559e303bc680c5f301944f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR