N-Carbobenzoxy-1,6-diaminohexane hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C1521-1G C1521-5G
TCC1521-001GEA 181.7 CAD
TCC1521-001G TCC1521-005G
N-Carbobenzoxy-1,6-diaminohexane hydrochloride ≥98.0% (by HPLC, total nitrogen)
N-Carbobenzoxy-1,6-diaminohexane hydrochloride

CAS Number: 78618-06-1
MDL Number: MFCD00270151
Molecular Formula: C14H22N2O2
Molecular Weight: 286.80
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 177


Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-(6-Aminohexyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl N-(6-Aminohexyl)carbamate Hydrochloride, N-Cbz-1,6-diaminohexane Hydrochloride

Formula: C₁₄H₂₂N₂O₂
Melting Pt: 177 °C
MDL Number: MFCD00270151
CAS Number: 78618-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a25ae0807e13b88be59a8909731c135f4f4c034a&vid=a25ae0807e13b88be59a8909731c135f4f4c034a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR