Methyl-5-chloro-2-methoxybenzoate ≥97.0%

Supplier: TCI America

Synonyms: 5-Chloro-2-methoxybenzoic acid methyl ester, Methyl 5-chloro-2-methoxybenzoate

C1457-25G
TCC1457-025GEA 363.4 CAD
TCC1457-025G
Methyl-5-chloro-2-methoxybenzoate ≥97.0%
Methyl 5-chloro-2-methoxybenzoate

CAS Number: 33924-48-0
MDL Number: MFCD00017511
Molecular Formula: C9H9ClO3
Molecular Weight: 200.62
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 113
Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: 5-Chloro-o-anisic Acid Methyl Ester, Methyl 5-Chloro-o-anisate, 5-Chloro-2-methoxybenzoic Acid Methyl Ester

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
Boiling Pt: 240 °C
Density: 1.28 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00017511
CAS Number: 33924-48-0
EINECS: 251-743-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=517213d195cf1613c60d9eb6e70fcbde99a4291c&vid=517213d195cf1613c60d9eb6e70fcbde99a4291c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR