(2-Carboxyethyl)dimethylsulfonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1241-5G
TCC1241-005GEA 219.62 CAD
TCC1241-005G
(2-Carboxyethyl)dimethylsulfonium bromide ≥98.0% (by titrimetric analysis)
(2-Carboxyethyl)dimethylsulfonium bromide

CAS Number: 20986-22-5
MDL Number: MFCD00142887
Molecular Formula: C5H11BrO2S
Molecular Weight: 215.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 112

Formula: C₅H₁₁BrO₂S
MW: 215.11 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00142887
CAS Number: 20986-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9557184f7e09fff0b36e4ed4c58e8b1801721703&vid=9557184f7e09fff0b36e4ed4c58e8b1801721703&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR