4-Carboxymethyl-6,7-methylenedioxycoumarin ≥95.0% (by titrimetric analysis)

Supplier: TCI America

C1196-1G
TCC1196-001GEA 355.34 CAD
TCC1196-001G
4-Carboxymethyl-6,7-methylenedioxycoumarin ≥95.0% (by titrimetric analysis)
4-Carboxymethyl-6,7-methylenedioxycoumarin

CAS Number: 97744-77-9
MDL Number: MFCD00143278
Molecular Formula: C12H8O6
Molecular Weight: 248.19
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 195

Formula: C₁₂H₈O₆
Melting Pt: 195 °C
MDL Number: MFCD00143278
CAS Number: 97744-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d7f864b5ee4234cd75155217d13e969511db761&vid=0d7f864b5ee4234cd75155217d13e969511db761&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR