2-Cyano-6-methoxybenzothiazole ≥97.0%

Supplier: TCI America

C1176-1G
TCC1176-001GEA 152 CAD
TCC1176-001G
2-Cyano-6-methoxybenzothiazole ≥97.0%
2-Cyano-6-methoxybenzothiazole

CAS Number: 943-03-3
MDL Number: MFCD00010537
Molecular Formula: C9H6N2OS
Molecular Weight: 190.22
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 130


Other Product Names / Synonyms: 6-Methoxy-2-benzothiazolecarbonitrile

Formula: C₉H₆N₂OS
Melting Pt: 130 °C
MDL Number: MFCD00010537
CAS Number: 943-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6fb02c77c6145423e8b16a80d032409f847bb8&vid=fd6fb02c77c6145423e8b16a80d032409f847bb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR