2-Chloro-6-ethoxypyridine ≥97.0%

Supplier: TCI America
Warning

C1143-25G
TCC1143-025GEA 237 CAD
TCC1143-025G
2-Chloro-6-ethoxypyridine ≥97.0%
2-Chloro-6-ethoxypyridine

CAS Number: 42144-78-5
MDL Number: MFCD00023465
Molecular Formula: C7H8ClNO
Molecular Weight: 157.60
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.17

Formula: C₇H₈ClNO
MW: 157.6 g/mol
Density: 1.17 (20 °C)
MDL Number: MFCD00023465
CAS Number: 42144-78-5
EINECS: 255-681-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1e1693caecc3ef46b94ee3ef6792d55784a85d9&vid=b1e1693caecc3ef46b94ee3ef6792d55784a85d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR