(-)-10-Camphorsulfonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0972-100G
TCC0972-100GEA 129.73 CAD
TCC0972-100G
(-)-10-Camphorsulfonic acid ≥98.0% (by titrimetric analysis)
L(-)-Camphorsulfonic acid

CAS Number: 35963-20-3
MDL Number: MFCD00064158
Molecular Formula: C10H16O4S
Molecular Weight: 232.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 195
Specific rotation [a]20/D: -21.5 deg (C=5, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Optical Resolving Agents, Bioscience, Reagents for Oligosaccharide Synthesis, Bicyclic Monoterpenes
Other Product Names / Synonyms: (-)-CSA

Formula: C₁₀H₁₆O₄S
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 195 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00150753
CAS Number: 35963-20-3
EINECS: 252-817-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c3a7acebaf055f623aa47ae81f12fdfdd75d15f&vid=6fdf6c8cb09fc7a185b7e2b1b3c760bba8089a66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR