6-Chloro-1-hexanol ≥96.0%

Supplier: TCI America

C0801-500G C0801-25G
TCC0801-500GEA 388.68 CAD
TCC0801-500G TCC0801-25G
6-Chloro-1-hexanol ≥96.0%
6-Chloro-1-hexanol

CAS Number: 2009-83-8
MDL Number: MFCD00002984
Molecular Formula: C6H13ClO
Molecular Weight: 136.62
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 98
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Hexamethylene Chlorohydrin

Formula: C₆H₁₃ClO
Boiling Pt: 117 °C
Density: 1.03 (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00002984
CAS Number: 2009-83-8
EINECS: 217-925-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dae77af8ccc4502bdadd7577ea46b27727db24b6&vid=254a2e4a85a2e0bb30471f37da4284d21c96451d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR