5-Chloro-2-methylaniline Hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-4-chlorotoluene Hydrochloride, 5-Chloro-2-methylaniline hydrochloride, 5-Chloro-o-toluidine Hydrochloride

C0746-25G
TCC0746-025GEA 243.32 CAD
TCC0746-025G
5-Chloro-2-methylaniline Hydrochloride ≥98.0%
5-Chloro-2-methylaniline Hydrochloride

CAS Number: 6259-42-3
MDL Number: MFCD00035487
Molecular Formula: C7H8ClN
Molecular Weight: 178.06
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Other Product Names / Synonyms: 2-Amino-4-chlorotoluene Hydrochloride, 5-Chloro-o-toluidine Hydrochloride

Formula: C₇H₈ClN
MDL Number: MFCD00035487
CAS Number: 6259-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2e8c6925cbf83a2c130cd33b81213e624c14a01&vid=e2e8c6925cbf83a2c130cd33b81213e624c14a01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR