trans-Cinnamic acid ≥99.8%, zone refined: number of passes 40

Supplier: TCI America
Warning

C0636-1SAMPLE
TCC0636-ZONEEA 453.46 CAD
TCC0636-ZONE
trans-Cinnamic acid ≥99.8%, zone refined: number of passes 40
trans-Cinnamic acid

CAS Number: 140-10-3
MDL Number: MFCD00004369
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >99.8% (GC)
Form: Crystal
Boiling point (°C): 300
Melting point (°C): 135
Lambda max.: 273 nm (MeOH)


Product Categories: Materials Chemistry, Highly Purified Reagents, Bioscience, Phenylpropanoids
Other Product Names / Synonyms: trans-3-Phenylacrylic Acid

Formula: C₉H₈O₂
Boiling Pt: 300 °C
Melting Pt: 135 °C
Density: 1.25 g/cm³ (4 °C)
MDL Number: MFCD00004369
CAS Number: 140-10-3
EINECS: 205-398-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9551c2b109a8a4b2477bcef256512688241cbe1e&vid=0d7768d96e8ba4139cc319a2414d17d868a23212&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.8% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR