Citrazinic acid ≥96.0% (by titrimetric analysis)

Supplier: TCI America

C0571-500G C0571-25G
TCC0571-500GEA 927.3 CAD
TCC0571-500G TCC0571-025G
Citrazinic acid ≥96.0% (by titrimetric analysis)
Citrazinic acid

CAS Number: 99-11-6
MDL Number: MFCD00006274
Molecular Formula: C6H5NO4
Molecular Weight: 155.11
Purity/Analysis Method: >96.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2,6-Dihydroxyisonicotinic Acid, 2,6-Dihydroxypyridine-4-carboxylic Acid

Formula: C₆H₅NO₄
MDL Number: MFCD00006274
CAS Number: 99-11-6
EINECS: 202-731-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15c6ebbd9a9c740ca51ee3ff3f7baaffc69dfb0a&vid=15c6ebbd9a9c740ca51ee3ff3f7baaffc69dfb0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR