L-Cystine dihydrochloride ≥98.0%

Supplier: TCI America

Synonyms: (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride, (2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid) dihydrochloride, H-Cystine-OH·2HCl

C0520-25G
TCC0520-025GEA 75.84 CAD
TCC0520-025G
L-Cystine dihydrochloride ≥98.0%
L-Cystine dihydrochloride

CAS Number: 30925-07-6
MDL Number: MFCD00070399
Molecular Formula: C6H12N2O4S2
Molecular Weight: 313.21
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -168 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: (H-Cys-OH)2.2HCl

Formula: C₆H₁₂N₂O₄S₂·2HCl
MDL Number: MFCD00070399
CAS Number: 30925-07-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56206bd31ec87c1b1e01d1d19f3be322488a88a8&vid=56206bd31ec87c1b1e01d1d19f3be322488a88a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR