trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C0473-500G C0473-25G C0473-100G
TCC0473-500GEA 1405.95 CAD
TCC0473-500G TCC0473-025G TCC0473-100G CA76097-940
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥99.0% (by titrimetric analysis)
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

CAS Number: 125572-95-4
MDL Number: MFCD00149243
Molecular Formula: C14H22N2O8
Molecular Weight: 346.34
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 216


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid, CyDTA

Formula: C₁₄H₂₂N₂O₈·1H₂O
Melting Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e050ef9e8ec5bc313b73496e3c38269baebaa2&vid=07e050ef9e8ec5bc313b73496e3c38269baebaa2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR