trans-4-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: p-Coumaric acid, trans-p-Cumaric Acid, trans-p-Coumaric Acid

C0393-500G C0393-25G C0393-100G
TCC0393-500GEA 813.23 CAD
TCC0393-500G TCC0393-025G TCC0393-100G
trans-4-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-4-Hydroxycinnamic acid

CAS Number: 501-98-4
MDL Number: MFCD00004399
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 213


Product Categories: Bioscience, Antioxidants, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: trans-p-Cumaric Acid, trans-4-Hydroxycinnamic Acid

Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 213 °C
MDL Number: MFCD00004399
CAS Number: 501-98-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5876702a5034ac1ddae38171322c8bb1217a2c02&vid=5876702a5034ac1ddae38171322c8bb1217a2c02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR