(Chloromethoxy)ethane ≥96.0% for synthesis

Supplier: TCI America

Synonyms: Chloromethyl ethyl ether

C0201-100G
TCC0201-100GEA 415.89 CAD
TCC0201-100G
(Chloromethoxy)ethane ≥96.0% for synthesis
(Chloromethoxy)ethane

CAS Number: 3188-13-4
MDL Number: MFCD00000887
Molecular Formula: C3H7ClO
Molecular Weight: 94.54
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 7
Specific Gravity (20/20): 1.02

Formula: ClCH₂OCH₂CH₃
Boiling Pt: 80 °C
Melting Pt: –98 °C
Density: 1.02 (20 °C)
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000887
CAS Number: 3188-13-4
EINECS: 221-687-5
UN: 2354
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cb737e98551ae005790ae1c446287f4ead64b4b&vid=f8883037e1ae6b03836ed88027f50b15fc0700cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR