Chlorohydroquinone ≥85.0%

Supplier: TCI America

Synonyms: 2-Chlorobenzene-1,4-diol, 2-Chlorohydroquinone

C0185-25G C0185-500G
TCC0185-025GEA 84.37 CAD
TCC0185-025G TCC0185-500G
Chlorohydroquinone ≥85.0%
Chlorohydroquinone

CAS Number: 615-67-8
MDL Number: MFCD00002341
Molecular Formula: C6H5ClO2
Molecular Weight: 144.55
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Boiling point (°C): 263
Melting point (°C): 105


Other Product Names / Synonyms: 1-Chloro-2,5-dihydroxybenzene

Formula: C₆H₅ClO₂
MW: 144.56 g/mol
Boiling Pt: 263 °C
Melting Pt: 105 °C
MDL Number: MFCD00002341
CAS Number: 615-67-8
EINECS: 210-442-8
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a2d441febbf682e13847f9fd1b5d0e0f4334462&vid=9a2d441febbf682e13847f9fd1b5d0e0f4334462&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR