5-Isopropyl-2-methylphenol ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Carvacrol, 5-Isopropyl-o-cresol, Isothymol, 5-Isopropyl-2-methylphenol

C0026-100G
TCC0026-100GEA 117.46 CAD
TCC0026-100G
5-Isopropyl-2-methylphenol ≥98.0% for synthesis
5-Isopropyl-2-methylphenol

CAS Number: 499-75-2
MDL Number: MFCD00002236
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 238
Flash Point (°C): 100
Specific Gravity (20/20): 0.98


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: Isothymol, 5-Isopropyl-o-cresol, 5-Isopropyl-2-methylphenol

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237
Melting Pt: 0.5
Density: 0.98 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00002236
CAS Number: 499-75-2
EINECS: 207-889-6
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e14352dca822e6f534da49e2ad2cbf1c3953e12c&vid=e14352dca822e6f534da49e2ad2cbf1c3953e12c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR