2-Bromo-3-hexadecylthiophene ≥96.0% (by GC)

Supplier: TCI America

B5409-1G
TCB5409-1GEA 292.3 CAD
TCB5409-1G
2-Bromo-3-hexadecylthiophene ≥96.0% (by GC)
2-Bromo-3-hexadecylthiophene
Formula: C₂₀H₃₅BrS
CAS Number: 827343-08-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR