3-Bromo-4'-iodo-1,1'-biphenyl ≥95.0% (by GC)

Supplier: TCI America

B5321-1G B5321-200MG
TCB5321-1GEA 325.48 CAD
TCB5321-1G TCB5321-200MG
3-Bromo-4'-iodo-1,1'-biphenyl ≥95.0% (by GC)
3-Bromo-4'-iodo-1,1'-biphenyl
Formula: C₁₂H₈BrI
MDL Number: MFCD28952954
CAS Number: 187275-73-6
UN: 3152
ADR: 9,II

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 95.0 %
Melting point 45.0 to 49.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR