Bosentan monohydrate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 4-(tert-Butyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]benzenesulfonamide, 4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-[2,2'-bipyrimidin]-4-yl)benzenesulfonamide hydrate, 4-tert-Butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidin-4-yl]benzenesulphonamide, 4-tert-Butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidin-4-yl]benzenesulfonamide, 4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-[2,2'-bipyrimidin]-4-yl)benzenesulphonamide hydrate, Bosentan hydrate, 4-(tert-Butyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]benzenesulphonamide

B5118-100MG B5118-25MG
TCB5118-100MGEA 263.86 CAD
TCB5118-100MG TCB5118-25MG
Bosentan monohydrate ≥98.0% (by HPLC)
Bosentan monohydrate
Formula: C₂₇H₂₉N₅O₆S·H₂O
MW: 551.62 g/mol
MDL Number: MFCD09751188
CAS Number: 157212-55-0
Merck Index: 14,01353

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.60 to 12.90 %
Water 2.5 to 4.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR